missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium, plasma standard solution, Specpure™ V 10,000μg/mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
| Physical Form | Liquid |
| Concentration | V2O5 in 5% HNO3 |
| UN Number | UN3264 |
| Quantity | 100 mL |
| Grade | Specpure™ |
| Chemical Name or Material | Vanadium plasma standard solution, V 10000μg/mL |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur