missing translation for 'onlineSavingsMsg'
Learn More

Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3

Product Code. 10480661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10480661 500 mL 500mL
10286630 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10480661 Supplier Thermo Scientific Acros Supplier No. 196435000
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    113.00 € / 500mL
    Save 9.00 €
    7% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-62-2 | V | 50.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanadium standard solution
Name Note 1 mg/ml V in 2% HNO3
Packaging Plastic bottle
CAS 7440-62-2, 7697-37-2, 7732-18-5
Physical Form Solution
Molecular Formula V
Concentration 995 to 1005 μg/mL of V(3+) in 2% HNO3 (20°C)
MDL Number MFCD00011453
Solubility Miscible with water
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES [V]
IUPAC Name vanadium
Molecular Weight (g/mol) 50.94
PubChem CID 23990
Quantity 500 mL
ChEBI CHEBI:35170
Formula Weight 50.94
Grade AAS
Show More Show Less
compliance-icons
Product Identifier
  • Vanadium standard solution, 1 mg/ml V(3+) in 2% HNO3
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.