missing translation for 'onlineSavingsMsg'
Learn More

Melford Vancomycin HCl

Product Code. p-200087157
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 30018919

Brand: Melford V065001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.

Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.

It is primarily effective against Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE).

In the field of microbiology and research, Vancomycin Hydrochloride is used as a selective agent in culture media. It is added to growth media to inhibit the growth of Gram-positive bacteria and selectively allow the growth of Gram-negative bacteria or other microorganisms. This property makes it a valuable tool in the isolation and identification of bacteria in laboratory settings.

Specifications

Chemical Name or Material Vancomycin HCl
CAS 1404-93-9
Quantity 1 g
Molecular Formula C66H75Cl2N9O24-HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.