missing translation for 'onlineSavingsMsg'
Learn More

Melford Vancomycin HCl

Product Code. 30018919
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018919 1 g 1g
30018921 5 g 5g
30018920 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018919 Supplier Melford Supplier No. V065001.0
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.

Vancomycin Hydrochloride is a widely used antibiotic that belongs to the glycopeptide class of antibiotics.

It is primarily effective against Gram-positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE).

In the field of microbiology and research, Vancomycin Hydrochloride is used as a selective agent in culture media. It is added to growth media to inhibit the growth of Gram-positive bacteria and selectively allow the growth of Gram-negative bacteria or other microorganisms. This property makes it a valuable tool in the isolation and identification of bacteria in laboratory settings.

Specifications

Chemical Name or Material Vancomycin HCl
CAS 1404-93-9
Quantity 1 g
Molecular Formula C66H75Cl2N9O24-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.