missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg

Product Code. 11956911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11956911 5 g Glass Bottle 5g
10379350 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11956911 Fournisseur Thermo Scientific Chemicals Code fournisseur 296990050
From Chemicals to Chromatography
Use promo code 27991
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Vancomycin hydrochloride (>900 microgram/mg), CAS # 1404-93-9, is an antibiotic from Streptomyces orientalis, that shows antibacterial activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic and antiviral from Streptomyces orientalis
  • It shows antibiotic activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains
  • Vancomycin hydrochloride inhibits bacterial mucopeptide synthesis

Applications

  • Vancomycin hydrochloride demonstrates potent activity against gram-positive bacteria
  • It has been shown in studies to be highly effective against methicillin-resistant Staphylococcus aureus (MRSA) in vitro and in vivo
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Spécification

Type Vancomycin Hydrochloride
Physical Form Powder
pH 2.5 to 4.5 (5% soln.)
Infrared Spectrum Authentic
Packaging Glass Bottle
Water 5% max.
Color Tan to White
Quantity 5 g
Solubility Information Solubility in water: 50g/L. Other solubilities: moderately soluble in diluted methanol, insoluble in higher alcohols, acetone and ether
Formula Weight 1485.73
Percent Purity ≥80.0% (vancomycin B) (HPLC), 9.0% (individual impurities) max., greater than 900μg/mg (potency, on anhydrous substance)
Chemical Name or Material Vancomycin hydrochloride
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Vancomycin hydrochloride
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.