missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Vancomycin ReadyMade™ Solution

Product Code. 30044049
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044049 5 x 10 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044049 Supplier TOKU-E™ Supplier No. V0405x10ML
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vancomycin ReadyMade™ Solution is provided as a sterile-filtered solution of Vancomycin Hydrochloride in DMSO.

  • CAS Number: 1404-93-9
  • Molecular Formula: C66H75Cl2N9O24 · HCl
  • Molecular Weight: 1485.71 g/mol
  • Storage Conditions: -20°C
  • Form: Solution
  • Source: Streptomyces orientalis
  • Spectrum: Vancomycin HCl inhibits growth of many Gram-positive bacteria including the antibiotic-resistant superbug, MRSA (methicillin-resistant Staphylococcus aureus). Vancomycin HCl is effective against MRSA because it inhibits cell wall synthesis through a different mechanism other than β-lactam antibiotics
  • Mechanism of Action: Vancomycin HCl prevents cell wall synthesis by two separate mechanisms. One mechanism prevents N-acetylmuramic acid (NAM) and N-acetylglucosamine (NAG) peptides from linking together forming the peptidoglycan backbone through the incorporation of the Vancomycin HCl molecule to the D-alanyl-D-alanine terminal. The second mechanism prevents crosslinking between amino acid residues in the peptidoglycan chain altering bacterial cell membrane permeability as well as RNA synthesis

Specifications

Chemical Name or Material Vancomycin Hydrochloride
Physical Form Liquid
CAS 1404-93-9
Source Streptomyces orientalis
Packaging Plastic bottle
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Colorless
Concentration 100 to 105 mg/mL
Quantity 5 x 10 mL
Purity Grade Notes >95% (HPLC)
Assay Percent Range >900 μg/mg
Assay Percent Notes Vancomycin Hydrochloride, Dried Basis
Molecular Formula C66H75Cl2N9O24 · HCl
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym Vancomycin Hydrochloride, Vancocine HCl, Vancocin HCl
IUPAC Name (1S,2R,18R,19R,22S,25R,28R,40S)-48-[(2S,3R,4S,5S,6R)-3-[(2S,4S,5S,6S)-4-amino-5-hydroxy-4,6-dimethyloxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-22-(2-amino-2-oxoethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[[(2R)-4-methyl-2-(methylamino)pentanoyl]amino]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentazaoctacyclo[26.14.2.23,6.214,17.18,12.129,33.010,25.034,39]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34(39),35,37,46,49-pentadecaene-40-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 1485.71 g/mol
Formula Weight 1485.71 amu
Percent Purity >95%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Vancomycin HCl 10%, Dimethyl Sulfoxide ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.