missing translation for 'onlineSavingsMsg'
Learn More

Vandetanib, MedChemExpress

Product Code. 16491546
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491546 500 mg 500mg
16490127 50 mg 50mg
16422846 25 mg 25mg
16404816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491546 Supplier Medchem Express Supplier No. HY10260
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vandetanib (D6474) is a potent, orally active inhibitor of VEGFR2/KDR tyrosine kinase activity (IC50=40 nM). Vandetanib also has activity versus the tyrosine kinase activity of VEGFR3/FLT4 (IC50=110 nM) and EGFR/HER1 (IC50=500 nM).

Vandetanib (D6474) is a potent, orally active inhibitor of VEGFR2/KDR tyrosine kinase activity (IC50=40 nM). Vandetanib also has activity versus the tyrosine kinase activity of VEGFR3/FLT4 (IC50=110 nM) and EGFR/HER1 (IC50=500 nM).

Specifications

Chemical Name or Material Vandetanib
CAS 443913-73-3
Color White
Purity Grade Notes Research
Molecular Formula C22H24BrFN4O2
Quantity 500 mg
Synonym ZD6474
Solubility Information DMSO : 27.5 mg/mL (57.85 mM; Need ultrasonic and warming)
SMILES FC1=CC(Br)=CC=C1NC2=NC=NC3=CC(OCC4CCN(CC4)C)=C(C=C23)OC
Molecular Weight (g/mol) 475.35
Formula Weight 475.35
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.