missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vang-like Protein 2/VANGL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 351.00 €
Specifications
| Antigen | Vang-like Protein 2/VANGL2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18170702
|
Novus Biologicals
NBP2-24704SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119941
|
Novus Biologicals
NBP2-24704 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vang-like Protein 2/VANGL2 Polyclonal specifically detects Vang-like Protein 2/VANGL2 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Vang-like Protein 2/VANGL2 | |
| Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 57216 | |
| A portion of amino acids 300-350 of human Van Gogh like-2 protein was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2 | |
| VANGL2 | |
| IgG | |
| Protein G purified | |
| 100% homologous in human isoform CRA_a, CRA_b and CRA_c). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title