missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, MedChemExpress

Product Code. 16481298
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
5 g
Unit Size:
200mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481298 5 g 5g
16421486 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481298 Supplier Medchem Express Supplier No. HYN0098
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vanillin (p-Vanillin) is a single molecule extracted from vanilla beans and also a popular odor used widely in perfume, food and medicine.

Vanillin (p-Vanillin) is a single molecule extracted from vanilla beans and also a popular odor used widely in perfume, food and medicine.

Specifications

Chemical Name or Material Vanillin
CAS 121-33-5
Color White
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 5 g
Synonym p-Vanillin m-Methoxy-p-hydroxybenzaldehyde p-Hydroxy-m-methoxybenzaldehyde
Solubility Information DMSO : 100 mg/mL (657.25 mM; Need ultrasonic)
SMILES O=CC1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.