missing translation for 'onlineSavingsMsg'
Learn More

Vanillyl alcohol, MedChemExpress

Product Code. 16459176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459176 Supplier Medchem Express Supplier No. HYN2067
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Vanillyl alcohol (p-(Hydroxymethyl)guaiacol), derived from vanillin, is a phenolic alcohol and is used as a flavoring agent in foods and beverages.

Vanillyl alcohol (p-(Hydroxymethyl)guaiacol), derived from vanillin, is a phenolic alcohol and is used as a flavoring agent in foods and beverages.

Specifications

Chemical Name or Material Vanillyl alcohol
CAS 498-00-0
Color White
Purity Grade Notes Research
Molecular Formula C8H10O3
Quantity 100 mg
Synonym p-(Hydroxymethyl)guaiacol
Solubility Information DMSO : 100 mg/mL (648.68 mM; Need ultrasonic)
SMILES OCC1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 154.16
Formula Weight 154.16
Percent Purity 98.38%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.