missing translation for 'onlineSavingsMsg'
Learn More

VAPA Mouse, anti-Human, Unconjugated, Clone: 1F10F9, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen VAPA
Clone 1F10F9
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17291506
View Documents
Proteintech
67845-1-IG-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17281506
View Documents
Proteintech
67845-1-IG-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.

Specifications

Specifications

VAPA
1000 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat
Primary
1F10F9
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Mouse
9218
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.