Learn More
Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (09) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-26914-20ug
 View more versions of this product
Product Code. 30028260
  • 218.00 € / 20µg
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Detection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Varicella Zoster Virus Glycoprotein E
Monoclonal
Unconjugated
0.2 μm filtered solution in PBS
Mouse
Protein A purified
RUO
Virus
Purified
ELISA, Western Blot, Sandwich ELISA Detection
09
Western Blot 0.2-1 μg/mL, ELISA 0.5-1 μg/mL, Sandwich ELISA Detection 0.1-1 μg/mL
Envelope glycoprotein E, GE, ORF68
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538).
20 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.