missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (17) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
218.00 € - 474.00 €
Specifications
| Antigen | Varicella Zoster Virus Glycoprotein E |
|---|---|
| Clone | 17 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043245
|
Novus Biologicals™
NBP3-26892-20ug |
20 μg |
218.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043969
|
Novus Biologicals™
NBP3-26892-100ug |
100 μg |
474.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Varicella Zoster Virus Glycoprotein E | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 17 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Envelope glycoprotein E, GE, ORF68 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title