missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Varicella zoster Virus glycoprotein E Recombinant Rabbit Monoclonal Antibody (276)
Rabbit Recombinant Monoclonal Antibody
169.00 € - 479.00 €
Specifications
| Antigen | Varicella zoster Virus glycoprotein E |
|---|---|
| Clone | 276 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30535922
|
Invitrogen™
MA560397 |
100 μg |
479.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535260
|
Invitrogen™
MA560398 |
20 μg |
169.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Varicella zoster Virus glycoprotein E Recombinant Monoclonal Antibody for Western Blot, ELISA
Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs predominantly in aging and immunosuppressed individuals' virons are spherical and 150-200 nm in diameter. Its lipid envelope encloses the nucleocapsid of 162 capsomeres arranged in a hexagonal form. Its DNA is a single linear, double strand molecule, 125,000 nt long.Specifications
| Varicella zoster Virus glycoprotein E | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Primary | |
| Protein A |
| 276 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| Recombinant Varicella-zoster virus (VZV)/HHV-3 gE/glycoprotein E protein (Met1-Tyr538). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title