missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Varicella zoster Virus (strain Dumas) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17298253 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17298253 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17298253 Supplier Invitrogen™ Supplier No. PA5115477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KITH_VZV7.

Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs precominantly in aging and immunosuppressed individuals. Glycoprotein E (gE), a 623 amino acid type I integral membrane protein, is expressed on plasma membranes and in the cytoplasm of VZV infected cells, where it is presumed to be present in the membranes of intracellular vesicles or organelles. VZV gE complexes with gI in the rough endoplasmic reticulum, and most gE exists in the form of gE to gI heterodimers in infected cells. VZV gE is also a predominant component of the virion envelope. The functions of gE, which is encoded by ORF68, are of particular interest because it has been demonstrated that ORF68 is essential for VZV replication. In contrast, the genes for gE proteins can be deleted from herpes simplex virus and pseudorabies virus, albeit with significant reductions in infectivity in cell culture and in animal models. Since the VZV genome does not encode a homologue of gD, VZV gE may have functions that are usually segregated between gD and gE, or the gE to gI complex, in other alphaherpesviruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Varicella zoster Virus (strain Dumas)
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Alias varicella zoster virus ge; Varicellovirus; VZV
Host Species Rabbit
Immunogen A synthesized peptide derived from human virus KITH_VZV7, corresponding to a region within the internal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.