missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP Antibody (GT238), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-13619
This item is not returnable.
View return policy
Description
VASP Monoclonal antibody specifically detects VASP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| VASP | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7408 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GT238 | |
| Western Blot, Immunohistochemistry, Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin | |
| vasodilator-stimulated phosphoprotein | |
| Recombinant protein encompassing a sequence within the C-terminus region of human VASP. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction