missing translation for 'onlineSavingsMsg'
Learn More

Vatalanib dihydrochloride, MedChemExpress

Product Code. 16491167
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491167 50 mg 50mg
16415706 100 mg 100mg
16404928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491167 Supplier Medchem Express Supplier No. HY12018
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM.

Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM.

Specifications

Chemical Name or Material Vatalanib dihydrochloride
Physical Form Solid
CAS 212141-51-0
Color Light Nude
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H17Cl3N4
For Use With (Application) Cancer-Kinase/protease
Synonym PTK787 dihydrochloride CGP-797870 dihydrochloride ZK-222584 dihydrochloride
Solubility Information DMSO : 50 mg/mL (119.12 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (119.12 mM; Need ultrasonic)
SMILES ClC1=CC=C(C=C1)NC2=NN=C(C3=C2C=CC=C3)CC4=CC=NC=C4.[H]Cl.[H]Cl
Molecular Weight (g/mol) 419.73
Formula Weight 419.73
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.