Learn More
Novus Biologicals™ VAV2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30790-250ug
 View more versions of this product
Product Code. 30067076
  • 508.00 € / 250µg
Description
VAV2 Polyclonal antibody specifically detects VAV2 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAV2
Polyclonal
Western Blot : 1:5000, Immunoprecipitation
guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2
A his tagged fusion protein VAV2 corresponding to amino acid sequence 578 - 878 and having a molecular weight of 100kD
250 μg
Signal Transduction
7410
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunoprecipitation, Western Blot
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Human
Purified