missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTCH2 Monoclonal Antibody (865619)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561909

Product Code. 30536248

  • 459.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTCH2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O35595
PTCH2
E. coli-derived recombinant mouse Patched 2/PTCH2, Glu79-Ser394.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunohistochemistry (Frozen)
865619
Unconjugated
PTCH2
patched 2; patched homolog 1; patched homolog 2; protein patched homolog 2; PTC2; Ptch1; PTCH2; RP11-269F19.8; UNQ560/PRO1121/PRO57079
Rat
Protein A/G
RUO
19207
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.