missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VCAM-1 Recombinant Rabbit Monoclonal Antibody (601), FITC
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560249
This item is not returnable.
View return policy
Description
VCAM-1 Recombinant Monoclonal Antibody for Flow
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
| VCAM-1 | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| P19320 | |
| VCAM1 | |
| Recombinant Human VCAM1 protein (Met1-Pro697). | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 601 | |
| FITC | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7412 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction