missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VCAM-1 Recombinant Rabbit Monoclonal Antibody (601), FITC
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

99.00 € - 209.00 €

Specifications

Antigen VCAM-1
Clone 601
Concentration 0.1 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30534253
GREENER_CHOICE
View Documents
Invitrogen™
MA560248
100 Tests
209.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30536247
View Documents
Invitrogen™
MA560249
25 Tests
99.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VCAM-1 Recombinant Monoclonal Antibody for Flow

VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications

Specifications

VCAM-1
0.1 mg/mL
Flow Cytometry
FITC
Rabbit
RUO
PBS with 0.5% BSA and 0.03% ProClin 300
P19320
7412
Recombinant Human VCAM1 protein (Met1-Pro697).
Antibody
601
4°C, store in dark, DO NOT FREEZE!
Recombinant Monoclonal
Liquid
IgG
Human
VCAM1
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
VCAM1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.