missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-68794
This item is not returnable.
View return policy
Description
VDAC2 Polyclonal specifically detects VDAC2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| VDAC2 | |
| Polyclonal | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| P45880 | |
| VDAC2 | |
| Peptide with sequence C-DSAKSKLTRNN corresponding to internal region according to NP_003366.2. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 7417 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction