missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VDP Recombinant Rabbit Monoclonal Antibody (24GB1905)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557834
This item is not returnable.
View return policy
Description
VDP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner.
Specifications
| VDP | |
| Recombinant Monoclonal | |
| Unconjugated | |
| USO1 | |
| 115kDa; General vesicular transport factor p115; P115; protein USO1 homolog; TAP; transcytosis associated protein; transcytosis associated protein p115; transcytosis-associated protein; USO1; USO1 homolog, vesicle docking protein; USO1 vesicle docking factor; USO1 vesicle docking protein homolog; USO1 vesicle transport factor; Vdp; VDP p115; vesicle docking protein p115; vesicle docking protein, 115 kDa; vesicle-docking protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56041, 56042, 8615 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1905 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60763, P41542, Q9Z1Z0 | |
| USO1 | |
| A synthesized peptide derived from human USO1 (aa 1-23). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction