Learn More
Invitrogen™ VDP Recombinant Rabbit Monoclonal Antibody (24GB1905)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557835
 View more versions of this product
Product Code. 30276450
  • 198.00 € / 20µL
Description
VDP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VDP
Recombinant Monoclonal
Unconjugated
USO1
115kDa; General vesicular transport factor p115; P115; protein USO1 homolog; TAP; transcytosis associated protein; transcytosis associated protein p115; transcytosis-associated protein; USO1; USO1 homolog, vesicle docking protein; USO1 vesicle docking factor; USO1 vesicle docking protein homolog; USO1 vesicle transport factor; Vdp; VDP p115; vesicle docking protein p115; vesicle docking protein, 115 kDa; vesicle-docking protein
Rabbit
Affinity chromatography
RUO
56041, 56042, 8615
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1905
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O60763, P41542, Q9Z1Z0
USO1
A synthesized peptide derived from human USO1 (aa 1-23).
20 μL
Primary
Human, Mouse, Rat