missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VE-cadherin Recombinant Rabbit Monoclonal Antibody (201), APC

Rabbit Recombinant Monoclonal Antibody
99.00 € - 209.00 €
Specifications
| Antigen | VE-cadherin |
|---|---|
| Clone | 201 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534185
![]() |
Invitrogen™
MA560413 |
100 Tests |
209.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535151
|
Invitrogen™
MA560414 |
25 Tests |
99.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-cadherin Recombinant Monoclonal Antibody for Flow
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.Specifications
| VE-cadherin | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P55284 | |
| 12562 | |
| Recombinant Mouse VE-Cadherin/CD144/CDH5 protein (Met1-Ala592). | |
| Antibody |
| 201 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| CDH5 | |
| 7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
| CDH5 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title