missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ VectaCell Trolox Antifade Reagent for Live Cell Imaging

Product Code. 18603200 Shop All Bio Techne Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
This item is not returnable. View return policy

Product Code. 18603200

Brand: Novus Biologicals™ CB10002NB

This item is not returnable. View return policy

Specifications

For Use With (Application) Immunocytochemistry/ Immunofluorescence, Cellular Imaging
Content And Storage Store at 4°C.
Quantity 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.