missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF164 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
211.00 € - 1855.00 €
Specifications
| Antigen | VEGF |
|---|---|
| Clone | 156902 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788548
![]() |
R&D Systems
MAB4931-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700588
![]() |
R&D Systems
MAB4931-100 |
100 μg |
459.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787139
![]() |
R&D Systems
MAB4931-500 |
500 μg |
1855.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal specifically detects VEGF in Mouse samples. It is validated for Western Blot.Specifications
| VEGF | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| VEGFA | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 156902 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| AAA40547 | |
| 7422 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGF 164 Ala27-Arg190 Accession # AAA40547 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse VEGF 164 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 25-100% cross-reactivity with recombinant human VEGF 165 and recombinant rat VEGF 164 and no cross-reactivity with recombinant mouse (rm) VEGF 115 rmVEGF 120 rmVEGF-B, rhVEGF-C, or rmVEGF-D. Cross-reactivity with rmVEGF 188 was not determined. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title