missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR2/KDR/Flk-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-529SS
This item is not returnable.
View return policy
Description
VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| VEGFR2/KDR/Flk-1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide | |
| KDR | |
| A synthetic peptide made to an internal region of the mouse VEGF Receptor 2 protein (between residues 1200-1300). [Swiss-Prot: P35918] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen sequence has 100% homology to rat. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 2.6 mg/mL | |
| Western Blot 1:250 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| Rabbit | |
| 150 kDa | |
| 0.025 mL | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| 3791 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction