Learn More
Invitrogen™ Venezuelan Equine Encephalitis Virus NSP4 Recombinant Rabbit Monoclonal Antibody (HL2023)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556700
Product Code. 30276678
  • 500.00 € / 100µL
Description
Venezuelan Equine Encephalitis Virus NSP4 Recombinant Monoclonal Antibody for Western Blot
Venezuelan Equine Encephalitis Virus (VEEV) is a mosquito-borne virus that can cause severe illness in both humans and animals. The virus is classified as a category B bioterrorism agent by the Centers for Disease Control and Prevention (CDC) due to its potential use as a biological weapon. The NSP4 gene of the VEEV is a non-structural protein that plays a crucial role in viral replication and pathogenesis. NSP4 is a promising target for the development of antiviral drugs and vaccines against VEEV.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Venezuelan Equine Encephalitis Virus NSP4
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Western Blot
HL2023
Unconjugated
VEEV NSP4
Recombinant fragment of Venezuelan equine encephalitis virus nsp4.
100 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles