missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-Verapamil hydrochloride, 99+%

Product Code. 10403045 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10403045 5 g Glass bottle 5g
10441012 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403045 Supplier Thermo Scientific Chemicals Supplier No. 329330050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H38N2O4·HCl
Molecular Weight (g/mol) 491.07
MDL Number MFCD00055208
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
PubChem CID 62969
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White
pH 4.5 to 6.5 (5% soln.)
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10144
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol
Formula Weight 491.07
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (±)-Verapamil hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (±)-Verapamil hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.