missing translation for 'onlineSavingsMsg'
Learn More

Veratrole, 99%

Product Code. 11444024
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444024 1000 g 1000g
11454024 250 g 250g
11464024 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444024 Supplier Thermo Scientific Alfa Aesar Supplier No. A11985.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-16-7 | C8H10O2 | 138.166 g/mol

It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-16-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00008357
InChI Key ABDKAPXRBAPSQN-UHFFFAOYSA-N
Synonym veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
PubChem CID 7043
ChEBI CHEBI:59114
IUPAC Name 1,2-dimethoxybenzene
SMILES COC1=CC=CC=C1OC

Specifications

Melting Point 21°C to 23°C
Density 1.084
Boiling Point 206°C to 207°C
Flash Point 87°C (188°F)
Refractive Index 1.533
Quantity 1000 g
Beilstein 1364621
Merck Index 14,9956
Solubility Information Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material Veratrole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.