missing translation for 'onlineSavingsMsg'
Learn More

Vernakalant Hydrochloride, TRC

Product Code. 30457244
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457244 5 mg 5mg
30455339 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457244 Supplier Toronto Research Chemicals Supplier No. TRCV1286205MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 748810-28-8
Molecular Formula C20H31NO4 . HCl
Molecular Weight (g/mol) 385.9
Synonym (3R)- 1-[(1R,2R)-2-[2-(3,4-Dimethoxyphenyl)ethoxy]cyclohexyl]-3-pyrrolidinol Hydrochloride, (1R,2R)-2-((3R)-Hydroxypyrrolidinyl)-1-(3,4-dimethoxyphenethoxy)cyclohexane Monohydrochloride, (1R,2R)-2-((3R)-Hydroxypyrrolidinyl)-1-[2-(3,4-dimethoxyphenyl)ethoxy]cyclohexane Monohydrochloride, (3R)-1-[(1R,2R)-2-[2-(3,4-Dimethoxyphenyl)ethoxy]cyclohexyl]-3-pyrrolidinol Monohydrochloride, RSD 1235
IUPAC Name (3R)-1-[(1R,2R)-2-[2-(3,4-dimethoxyphenyl)ethoxy]cyclohexyl]pyrrolidin-3-ol;hydrochloride
SMILES Cl.COc1ccc(CCO[C@@H]2CCCC[C@H]2N3CC[C@@H](O)C3)cc1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H31NO4.ClH/c1-23-19-8-7-15(13-20(19)24-2)10-12-25-18-6-4-3-5-17(18)21-11-9-16(22)14-21;/h7-8,13,16-18,22H,3-6,9-12,14H2,1-2H3;1H/t16-,17-,18-;/m1./s1
Formula Weight 385.202
Chemical Name or Material Vernakalant Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.