Learn More
3M™ Versaflo™ Battery, High Capacity
Product Code. 18899644 Shop All 3M Products
Change view
Click to view available options
Model:
TR-332
TR-630
TR-632
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
18899644 TR-632 Each
30305880 TR-332 Each
30305882 TR-630 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18899644 Supplier 3M™ Supplier No. 7100327727
  • ONLINE EXCLUSIVE PRICE
    731.40 €
    565.93 € / Pack of 1
    Save 165.47 €
    22% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Powered Air Respirator Batteries are for use with specific powered air respirators.
  • Available in multiple options : standard capacity, high capacity, and intrinsically safe.
  • Compatible with select powered air respirator systems.
  • Designed to support various user needs and safety requirements.
  • Ensures reliable power for extended usage durations.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Product Type Battery
Model TR-632