Learn More
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units
Product Code. 10099369 Shop All 3M Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10099369 Small/Medium Each
10566490 Medium/Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10099369 Supplier 3M™ Supplier No. 7000000158
  • ONLINE EXCLUSIVE PRICE
    91.00 €
    63.63 € / Pack of 1
    Save 27.37 €
    30% Off
View all promotions
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Lightweight BT-series breathing tubes are compatible with all 3M™ Versaflo™ headtops and air delivery units.
  • Long, lightweight breathing tube that is easy to wipe down
  • Quick release swivel enables one-handed connection and release
  • Disposable breathing tube cover is available to help cleaning and decontamination operations
  • 3M Catalog# BT-20S and BT-20L
TRUSTED_SUSTAINABILITY
Specifications
Product Type Breathing Tube
Size Small/Medium
For Use With (Application) Assembly, facility maintenance, grinding, machine operation, painting, sanding
For Use With (Equipment) Versaflo™ PAPR Systems
Model BT-20S
Length (English) 28 in.
Length (Metric) 71.1 cm