Learn More
3M™ Versaflo™ Industrial Helmet with Hood Powered Air Respirator Kit, TR-600-IHK
Product Code. 31297043 Shop All 3M Products
Change view
Click to view available options
Size:
One Size
Product Code. Size
31297043 One Size
1 options
Product Code. 31297043 Supplier 3M™ Supplier No. 7100317976
  • ONLINE EXCLUSIVE PRICE
    2970.00 €
    1844.69 € / Pack of 1
    Save 1125.31 €
    37% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Versatile and easy-to-use respiratory protection against certain gases and vapours and as well as particulates.
  • TR-600 Powered Air Respirator Systems are engineered for versatility and customisation and offer a variety of headtops, breathing tubes, and accessories to create a customised Powered Air Respirator solution.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Product Type PAPR Assembly with Hood
Size One Size