Learn More
3M™ Versaflo™ Mining Lamp Bracket Kit M-940, for M-Series Hard Hats and Helmets
Product Code. 18634333 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
18634333 1 Ea. Each
1 options
Product Code. 18634333 Supplier 3M™ Supplier No. 7100265324
  • ONLINE EXCLUSIVE PRICE
    135.00 €
    91.72 € / Pack of 1
    Save 43.28 €
    32% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The 3M™ Versaflo™ Mining Lamp Bracket, M-940, is designed to securely hold a range of fixed and adjustable angle mining lamps. It is compatible with certain M-series helmets and is designed for use with 3M™ Versaflo™ Powered Air Respirators. The bracket is made of a lightweight durable polymer and is cleanable and easy to install.
  • The bracket is made of a lightweight durable polymer
  • Can be used with a range of corded and cordless mining lamps
  • The bracket is easy to install and easy to adjust for a desired lamp angle
  • Designed to allow the user to lift the visor without removing mining lamp
  • The design helps hold mining lamps securely
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Material Plastic
Product Type Mining Lamp Bracket
Size One Size
Quantity 1 Ea.