missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Powered Air Respirator System Easy Clean Ready Kit, TR-300E+ ECK

Product Code. 17798478 Shop All 3M Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17798478 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17798478 Supplier 3M™ Supplier No. 7100233764
  • ONLINE EXCLUSIVE PRICE
    1805.00 €
    1261.37 € / Pack of 1
    Save 543.63 €
    30% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Versaflo™ TR-300+ Powered Air Respirators are versatile and easy-to-use respiratory protection against particulates hazards.

  • Slim, lightweight unit is well-balanced and comfortable to wear for long work shifts
  • Two user selectable airflows enable the user to control flow settings to allow the user to find the setting they find most comfortable
  • Powered Air Respirators automatically adjust airflow to compensate for filter loading to deliver consistent filtered air during use
  • Audible and visual alerts for low airflow and low battery charge-level to keep you prepared and informed
  • Able to be worn on a belt or backpack
  • Available in convienient, easy to buy assemblies and kits
TRUSTED_SUSTAINABILITY

Specifications

Battery Life 9 to 12 hr.
Battery Type High Capacity Lithium Ion - Rechargeable
Product Type PAPR System for Heavy Industry
For Use With (Equipment) TR-3712E Filter
Includes S-655 Hood, BT-30 Length Adjusting Breathing Tube, TR-302E+ Powered Air Turbo, TR-327 Easy Clean Belt, TR-332 High Capacity Battery, TR-341E Battery Charger Kit, TR-3712E (P) Filter, TR-3600 Pre-Filter, TR-971 Airflow Indicator
Model TR-300E+ ECK
Quantity 1
Dimensions (L x W x H) 60.0 x 29.0 x 23.0 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.