Learn More
3M™ Versaflo™ Premium S-Series Hood and Hard Hat Adapter Assembly, S-960
Product Code. 31297041 Shop All 3M Products
Change view
Click to view available options
Product Type:
Hood and Hard Hat Adapter Assembly
Product Code. Product Type
31297041 Hood and Hard Hat Adapter Assembly
1 options
Product Code. 31297041 Supplier 3M™ Supplier No. 7100308819
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    83.89 € / Pack of 1
    Save 48.11 €
    36% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to fit the 3M™ SecureFit™ Hard Hat H-700 Series and the 3M™ Hard Hat G3000 Series.
  • Compatible with 3M™ Versaflo™ S-Series Premium Hoods and 3M™ Versaflo™ Powered and Supplied Air Respirator Systems.
  • Provides integrated head, face, shoulder, and respiratory protection from dust and spray.
  • Helps direct airflow to the front of the hood for improved comfort and visibility.
  • Enables easy integration of head and respiratory protection for enhanced worker safety.
  • Lightweight and durable design for comfortable, long-term use in industrial environments.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Single-use
Material Plastic
Product Type Hood and Hard Hat Adapter Assembly
Size One Size