missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Radiant Heat Cover

Product Code. 17247640
Change view
Click to view available options
For Use With (Equipment):
3 M™ Versaflo™ Helmet M-307
3 M™ Versaflo™ Helmet M-407
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17247640 3 M™ Versaflo™ Helmet M-307 Each
17257640 3 M™ Versaflo™ Helmet M-407 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17247640 Supplier 3M™ Supplier No. 7100099853
  • ONLINE EXCLUSIVE PRICE
    713.00 €
    506.29 € / Pack of 1
    Save 206.71 €
    28% Off
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Made from lightweight, aluminised aramid fabric and provides improved head, neck and shoulder protection and comfort from radiant heat

  • Extend M-Series use in high temperature environments by providing improved head, neck and shoulder protection
  • Lightweight aluminised aramid fabric
  • CE Approved
TRUSTED_SUSTAINABILITY

Specifications

Description M-973
Color Gray
For Use With (Equipment) 3 M™ Versaflo™ Helmet M-307
Length (Metric) 0.355 m
Width (Metric) 0.228 m
Height (Metric) 0.086 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.