missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ M-300 Series Respiratory Helmet

Product Code. 11763916
Change view
Click to view available options
Versaflo™ M-300 Series Respiratory Helmet; Model: M-306; Includes: Comfort Faceseal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11763916 Versaflo™ M-300 Series Respiratory Helmet; Model: M-306; Includes: Comfort Faceseal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11763916 Supplier 3M™ Supplier No. 7000104035

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Versaflo™ M-300 Series Faceshields are lightweight and compact. Supplied with various faceseal options they feature an adjustable textile harness and airflow deflector, and can be used with 3M™ Air Delivery Units. Head protection to EN397 allows construction site use.

Features:

  • Chemical and Scratch Resistant Lenses
  • Large Viewing Area
  • Lightweight
  • Easy Visor Replacement System

Environmental Conditions:

  • Cold Working Environment
  • High Mobility Required
  • Safety Helmet Required

Hazard Type:

  • Gas, Vapour and Particulates
  • Liquid Splash
  • Low Energy Flying Particles/Dust
  • Falling Objects
  • Sparks and Hot Particles
TRUSTED_SUSTAINABILITY

Especificaciones

Assigned Protection Factor (APF) NPF: 200 (V-Series), NPF: 50 (Jupiter), NPF: 500 (TR-302E/TR-602E)
Certifications/Compliance CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001, EN 397
Color Black
Material Polyurethane-coated Polyamide
Product Type Respirator Helmet
For Use With (Application) Demolition, Food and Beverage Manufacturing, Metalworking, Pulp and Paper, Agriculture, Smelting, Woodworking, Chemical Processing, Construction
For Use With (Equipment) 3M Air Delivery Units
Includes Comfort Faceseal
Material (Lens) Coated polycarbonate
Model M-306
Quantity 1 Ea.
Sterility Non-sterile
Suspension Type Textile
Visor Type Clear
With Faceseal Yes
Dimensions (L x W x H) 346 x 346 x 254 mm
Length (English) 13.6 in.
Length (Metric) 346 mm
Width (English) 13.6 in.
Width (Metric) 346 mm
Height (English) 9.6 in.
Height (Metric) 254 mm
Weight (English) 1.8 lb.
Weight (Metric) 832 g
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.