missing translation for 'onlineSavingsMsg'
Learn More

Vesicular Stomatitis Indiana Virus Glycoprotein Antibody (110) - (San Juan), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-40579-20ul

 View more versions of this product

Product Code. 30238495

  • 214.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Vesicular Stomatitis Indiana Virus Glycoprotein Monoclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vesicular Stomatitis Indiana Virus Glycoprotein
Monoclonal
Unconjugated
0.2 μm filtered solution in PBS
This antibody was obtained from a rabbit immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein - (San Juan) (RefSeq#: NP_041715.1; Met1-Ser467). HEK293 used as expression system.
20 μL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
ELISA
110
ELISA 1:5000-1:10000
Rabbit
Protein A purified
RUO
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.