missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VG5Q Monoclonal Antibody (368011)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561250

Product Code. 30535548

  • 429.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

VG5Q (Aggf1) gene encodes an angiogenic factor that promotes proliferation of endothelial cells. Mutations in this gene are associated with a susceptibility to Klippel-Trenaunay syndrome. Pseudogenes of this gene are found on chromosomes 3, 4, 10 and 16.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VG5Q
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q7TN31
AGGF1
E. coli-derived recombinant mouse VG5Q, Met1-Glu711.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunocytochemistry
368011
Unconjugated
AGGF1
2010009L17Rik; 2310029P06Rik; AGGF1; Angiogenic factor VG5Q; angiogenic factor with G patch and FHA domains 1; angiogenic factor with G-patch and FHA domains 1; AW112072; FLJ10283; G patch domain-containing protein 7; GPATC7; GPATCH7; HSU84971; HUS84971; hVG5Q; mVG5Q; Peg3; RGD1310888; Vasculogenesis gene on 5q protein; Vg5q
Rat
Protein A/G
RUO
66549
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.