Learn More
Invitrogen™ VGLUT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 482400
Description
This antibody is specific for the VGLUT1 protein. On western blots, it identifies the target band at ~55 kDa. Reactivity has been confirmed with mouse brain homogenates by western blotting, with mouse hippocampus frozen sections by immunohistochemistry, and in immunofluorescence analysis with human glutamate neurons derived from iPSCs.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
Specifications
| VGLUT1 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A7 | |
| 2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116638, 57030, 72961 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Q3TXX4, Q62634, Q9P2U7 | |
| SLC17A7 | |
| Synthetic peptide derived from the internal region of the mouse VGLUT1 protein (Accession# NP_892038, Q3TXX4), which is 93% homologous to human and rat. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.