missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT2 Antibody (CL2952) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46641AF647
This item is not returnable.
View return policy
Description
VGLUT2 Monoclonal antibody specifically detects VGLUT2 in Human,Mouse,Rat,Macaque samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
Specifications
| VGLUT2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2 | |
| This antibody was developed using a recombinant protein derived from Q9P2U8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 57084 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CL2952 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction