missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIAAT/SLC32A1/VGAT Antibody (CL2793) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46628AF700
This item is not returnable.
View return policy
Description
VIAAT/SLC32A1/VGAT Monoclonal antibody specifically detects VIAAT/SLC32A1/VGAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| VIAAT/SLC32A1/VGAT | |
| Monoclonal | |
| Alexa Fluor 700 | |
| bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT | |
| This antibody was developed using a recombinant protein derived from Q9H598, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience, Neurotransmission | |
| 140679 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL2793 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction