Learn More
Epredia™ Lab Vision™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody
Product Code. 12692167 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Unit Size:
100µL
12mL
1mL
500µL
7mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12692167 7 mL Prediluted 7mL
12622177 100 μL Concentrated 100µL
12632177 500 μL Concentrated 500µL
12612177 1 mL Concentrated 1mL
12602177 12 mL Ready-to-Use 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12692167 Supplier Epredia™ Supplier No. MS728R7
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody.
CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.
Host Species: Mouse
Clone: 56C6
Isotype: IgG1
Species Reactivity: Human and Rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD10 protein
Molecular Weight: 100kDa
Positive Control: Small intestine
Cellular Localization: Membrane
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigen CD10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 56C6
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant external domain of CD10 protein
Quantity 7 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Rat
Form Prediluted
Isotype IgG1