Learn More
Gibco™ Penicillin-Streptomycin (10,000 U/mL)
Product Code. 15276355
Change view
Click to view available options
Quantity:
20 mL
100 mL
20 x 100 mL
Unit Size:
100mL
20mL
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15276355 20 x 100 mL Pack of 20
11568876 20 mL 20mL
11548876 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15276355 Supplier Gibco™ Supplier No. 15140163
For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.
  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Concentration 100 X
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Product Line Gibco
Quantity 20 x 100 mL
My Penicillin-Streptomycin solution is not colorless. Is this normal?
Yes, this is normal and will not affect the potency or application of the product. This solution is typically colorless. However, it can have a pink to yellow color tint. The coloring is a carry-over from the manufacturing process of Streptomycin - the genus that Steptomycin is isolated from (Actinomycetes Streptomyces griseus) is responsible for a wide variety of pigments.
How can I decontaminate my cultures?
When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.
The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.
What antibiotics do you offer to help control or eliminate cell culture contamination?
Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).
For Research Use Only. Not for use in diagnostic procedures.