Learn More
Invitrogen™ AMDHD1 Polyclonal Antibody
GREENER_CHOICE
Product Code. 15848303
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15848303 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15848303 Supplier Invitrogen™ Supplier No. PA569894
Rabbit Polyclonal Antibody
This target displays homology in the following species: Cow: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 92%; Rabbit: 79%; Rat: 86%.
AMDHD1 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Antigen AMDHD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AMDHD1
Gene Accession No. Q96NU7
Gene Alias 1300019J08Rik; Amdhd1; amidohydrolase domain containing 1; amidohydrolase domain-containing protein 1; HMFT1272; Probable imidazolonepropionase; RGD1308879
Gene Symbols AMDHD1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human AMDHD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144193
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG