Learn More
NFATC2IP, Human, IP-WB Antibody Pair (PW3), Abnova™
Description
Includes
Antibody pair for IP: rabbit polyclonal anti-NFATC2IP (300μL), Antibody pair for WB: mouse purified polyclonal anti-NFATC2IP (50μg)
Specifications
Specifications
| Antigen | NFATC2IP |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | NFATC2IP |
| Gene Alias | FLJ14639/MGC126790/MGC138387 |
| Gene Symbols | NFATC2IP |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |