Learn More
Ivermectin, MedChemExpress
Product Code. 16471499
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471499 1 g 1g
16471766 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16471499 Supplier Medchem Express Supplier No. HY15310
MedChemExpress Ivermectin (MK-933) is a broad-spectrum anti-parasite agent.
Ivermectin (MK-933) is a specific inhibitor of Impα/β1-mediated nuclear import and has potent antiviral activity towards both HIV-1 and dengue virus. It is a positive allosteric effector of P2X4 and the α7 neuronal nicotinic acetylcholine receptor (nAChRs). Ivermectin also inhibits bovine herpesvirus1 (BoHV-1) replication and inhibits BoHV-1 DNA polymerase nuclear import. Ivermectin is a candidate therapeutic against SARS-CoV-2/COVID-19.
Specifications
Chemical Name or Material Ivermectin
Physical Form Solid
CAS 70288-86-7
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C48H74O14
For Use With (Application) COVID-19-anti-virus
Synonym MK-933
Solubility Information DMSO : 250 mg/mL (285.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@@H]3C[C@H]4C[C@@]5(O[C@]([H])(C(C)C)[C@@H](C)CC5)O3)[C@@H](C)/C=C/C=C6CO[C@@]7([H])[C@]\6(O)[C@H](C(O4)=O)C=C(C)[C@H]7O)C[C@@H]2OC)O[C@@H](C)[C@@H]1O.CO[C@H]8C[C@H](O[C@H]9[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@@H]%10C[C@H]%11C[C@@]%12(O[C@]([H])([C@@H](C)CC)[C@@H](C)CC%12)O%10)[C@@H](C)/C=C/C=C%13CO[C@@]%14([H])[C@]\%13(O)[C@H](C(O%11)=O)C=C(C)[C@H]%14O)C[C@@H]9OC)O[C@@H](C)[C@@H]8O
Molecular Weight (g/mol) 875.09
Formula Weight 875.09
Percent Purity 96.68%
Grade Research